missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine Monohydrate 98.0+%, TCI America™

Catalog No. M00635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00635G 5 g
M00631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M00635G Supplier TCI America Supplier No. M00635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6112-76-1
Molecular Formula C5H6N4OS
Molecular Weight (g/mol) 170.19
MDL Number MFCD03854445,MFCD01461928
InChI Key WFFQYWAAEWLHJC-UHFFFAOYSA-N
Synonym 6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5
PubChem CID 2724350
ChEBI CHEBI:31822
IUPAC Name 6,7-dihydro-3H-purine-6-thione hydrate
SMILES O.S=C1N=CNC2=C1NC=N2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 152.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Mercaptopurine Monohydrate
EINECSNumber (9)-1432
RTECSNumber UP0400000
TSCA No