missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine monohydrate, 98%

Catalog No. AC226520250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226520250 25 g Glass bottle
AC226520050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226520250 Supplier Thermo Scientific Chemicals Supplier No. 226520250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6112-76-1 | C5H6N4OS | 170.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6112-76-1
Molecular Formula C5H6N4OS
Molecular Weight (g/mol) 170.19
MDL Number MFCD03854445,MFCD01461928
InChI Key WFFQYWAAEWLHJC-UHFFFAOYSA-N
Synonym 6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5
PubChem CID 2724350
ChEBI CHEBI:31822
IUPAC Name 3,7-dihydropurine-6-thione;hydrate
SMILES O.S=C1N=CNC2=C1NC=N2

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 313.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 10, 5702
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted alkali solutions,slightly soluble in diluted sulfuric acid,soluble in hot alcohol,insoluble in acetone and ether
Formula Weight 170.19
Percent Purity 99.5+%
Physical Form Fine Powder
Chemical Name or Material 6-Mercaptopurine monohydrate, 99.5+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
RTECSNumber UP0400000

RUO – Research Use Only