accessibility menu, dialog, popup

UserName

CAS RN 61078-14-6

IUPAC Name: 2-methanesulfonyl-1-methyl-1H-1,3-benzodiazole
Synonyms: 2-methanesulfonyl-1-methyl-1,3-benzodiazole
Molecular Weight (g/mol): 210.25
Molecular Formula: C9H10N2O2S
InChi Key: VDZVTXFWEKBHBV-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2N=C1S(C)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results