accessibility menu, dialog, popup

UserName

CAS RN 60804-75-3

NNNNNNFPFFFFFFPFFFFFRu

CAS RN 60804-75-3

IUPAC Name: λ²-ruthenium(2+) tris(1,10-phenanthroline) bis(hexafluoro-λ⁵-phosphanuide)
Synonyms: λ²-ruthenium(2+) tris(phen) dihexafluorophosphate
Molecular Weight (g/mol): 931.63
Molecular Formula: C36H24F12N6P2Ru
InChi Key: YRYUXGTVQZIGNQ-UHFFFAOYSA-N
SMILES: [Ru++].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results