missing translation for 'onlineSavingsMsg'
Learn More

Lead Subacetate TS, Ricca Chemical

Catalog No. R4312000120
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4312000120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4312000120 Supplier Ricca Chemical Company Supplier No. R4312000120C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6080-56-4 , 1317-36-8 , 7732-18-5
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless to slight acetate odor
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 120 mL
Chemical Name or Material Lead Subacetate TS