missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate TS, 9.5% (w/v), Ricca Chemical

Catalog No. 429316
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
429316 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 429316 Supplier Ricca Chemical Company Supplier No. 429316

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meets USP Specifications

Chemical Identifiers

CAS 6080-56-4 , 7732-18-5
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.333
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name lead(2+);diacetate;trihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]

Specifications

CAS Min % 89.0, 8.51
CAS Max % 93.0, 8.85
Grade USP
Packaging Natural Poly Bottle
Quantity 500 mL
Chemical Name or Material LEAD ACETATE TS (USP)