accessibility menu, dialog, popup

UserName

H3CH3CCH3PCH3CH3H3CCH3CH3H3CH3CH3CCH3PCH3CH3H3CCH3CH3H3CPt

CAS RN 60648-70-6

IUPAC Name: bis(tri-tert-butylphosphane) platinum
Synonyms: bis(tri-tert-butylphosphane) platinum
Molecular Weight (g/mol): 599.73
Molecular Formula: C24H54P2Pt
InChi Key: RJQWVEJVXWLMRE-UHFFFAOYSA-N
SMILES: [Pt].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results