missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-tert-butylphosphine)platinum(0), TCI America™

Catalog No. B3162250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3162250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3162250MG Supplier TCI America Supplier No. B3162250MG
Only null left

Chemical Identifiers

CAS 60648-70-6
Molecular Formula C24H54P2Pt
Molecular Weight (g/mol) 599.728
MDL Number MFCD04974248
InChI Key RJQWVEJVXWLMRE-UHFFFAOYSA-N
PubChem CID 6097049
IUPAC Name platinum;tritert-butylphosphane
SMILES CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C.[Pt]

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 250 mg
Formula Weight 599.73
Physical Form Crystalline Powder
Chemical Name or Material Bis(tri-tert-butylphosphine)platinum(0)