CAS RN 60407-35-4
CAS RN 60407-35-4
IUPAC Name:
1-benzylpiperidin-3-amine
Synonyms:
1-benzylpiperidin-3-amine
Molecular Weight (g/mol):
190.29
Molecular Formula:
C12H18N2
InChi Key:
HARWNWOLWMTQCC-UHFFFAOYNA-N
SMILES:
NC1CCCN(CC2=CC=CC=C2)C1
1
–
1
of
1
results
1
3-Amino-1-benzylpiperidine 98.0+%, TCI America™
CAS: 60407-35-4 Molecular Formula: C12H18N2 Molecular Weight (g/mol): 190.29 MDL Number: MFCD04038444 InChI Key: HARWNWOLWMTQCC-UHFFFAOYSA-N PubChem CID: 421694 IUPAC Name: 1-benzylpiperidin-3-amine SMILES: C1CC(CN(C1)CC2=CC=CC=C2)N