accessibility menu, dialog, popup

UserName

CAS RN 602-25-5

OOClCl

CAS RN 602-25-5

IUPAC Name: 1,4-dichloro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,4-dichloroanthracene-9,10-dione
Molecular Weight (g/mol): 277.1
Molecular Formula: C14H6Cl2O2
InChi Key: CAHGWVAXFJXDNI-UHFFFAOYSA-N
SMILES: ClC1=C2C(=O)C3=CC=CC=C3C(=O)C2=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results