missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dichloroanthraquinone 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 190°C |
| Quantity | 5 g |
| Formula Weight | 277.10 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 1,4-Dichloroanthraquinone |
Sécurité et manipulation
| TSCA | No |