accessibility menu, dialog, popup

UserName

CAS RN 60-92-4

IUPAC Name: (4aR,6R,7R,7aS)-6-(6-amino-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Synonyms: cyclic adenosine monophosphate
Molecular Weight (g/mol): 329.21
Molecular Formula: C10H12N5O6P
InChi Key: IVOMOUWHDPKRLL-YPLCUDRINA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)