missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 3',5'-Cyclic Monophosphate Hydrate 99.0+%, TCI America™

Catalog No. A23815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23815G 5 g
A23811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23815G Supplier TCI America Supplier No. A23815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-92-4
Molecular Formula C10H12N5O6P
Molecular Weight (g/mol) 329.21
MDL Number MFCD00005845
InChI Key IVOMOUWHDPKRLL-YPLCUDRINA-N
Synonym camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
PubChem CID 6076
ChEBI CHEBI:17489
IUPAC Name (4aR,6R,7R,7aS)-6-(6-amino-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
SMILES NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Melting Point 220°C
Color White
Quantity 5 g
Formula Weight 329.21
Percent Purity ≥99.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Adenosine 3′,5′-Cyclic Monophosphate Hydrate
EINECSNumber (9)-0044
RTECSNumber AU7357600
TSCA Yes
Recommended Storage Freezer