accessibility menu, dialog, popup

UserName

OH

CAS RN 60-12-8

IUPAC Name: 2-phenylethan-1-ol
Synonyms: 2-phenyl-ethanol phenethanol fema number 2858 epa pesticide chemical code 001503 β-PEA β-fenethylalkohol phenyl ethyl alcohol (substance) β-phenethyl alcohol phenethyl alcohol (natural) caswell no. 655C
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: WRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC=C1
Boiling Point 
  • (8)
  • (5)
  • (6)
Physical Form 
  • (11)
  • (8)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (8)
  • (8)
  • (2)
  • (9)
  • (8)
  • (5)
  • (6)
  • (11)
  • (8)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (6)
  • (4)
  • (4)