missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl Alcohol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014828
Change view
Click to view available options
Quantity:
2 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014828 2 g
111014827 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014828 Supplier Merck Emd Millipore Supplier No. PHR11222G
Only null left
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 60-12-8
Vapor Pressure 1 mmHg (58°C)
Vapor Density 4.21 (vs. air)
Density 1.020 g/mL (at 20°C)
Flash Point 102°C - Closed Cup
Physical Form Neat
Linear Formula C6H5 CH2 CH2 OH
MDL Number MFCD00002886
Melting Point -27°C
Boiling Point 219°C to 221°C (750 mmHg)
Synonym 2-Phenylethanol; beta-PEA; 2-Phenylethyl alcohol; Benzyl carbinol; PEA
Type Certified Reference Material
Quantity 2 g
Formula Weight 122.16
Grade Certified Reference Material
Show More Show Less