CAS RN 599-94-0
IUPAC Name:
[2-(benzenesulfonyl)ethanesulfonyl]benzene
Synonyms:
[2-(benzenesulfonyl)ethanesulfonyl]benzene
Molecular Weight (g/mol):
310.38
Molecular Formula:
C14H14O4S2
InChi Key:
ULELOBVZIKJPAC-UHFFFAOYSA-N
SMILES:
O=S(=O)(CCS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1,2-Bis(phenylsulfonyl)ethane, 98+%
1,2-Bis(phenylsulfonyl)ethane, 98+%
CAS: 599-94-0 Molecular Formula: C14H14O4S2 Molecular Weight (g/mol): 310.38 MDL Number: MFCD00041256 InChI Key: ULELOBVZIKJPAC-UHFFFAOYSA-N Synonym: 1,2-bis phenylsulfonyl ethane,1,2-bis phenylsulphonyl ethane,1,1'-ethane-1,2-diylbis sulphonyl bisbenzene,2-benzenesulfonyl ethylsulfonylbenzene,ethylenebisphenyl sulfone,2-phenylsulfonyl ethyl sulfonyl benzene,2-benzenesulfonyl ethanesulfonyl benzene,1,1'-ethane-1,2-diyldisulfonyl dibenzene,ethylene diphenylsulfone,acmc-1apw3 PubChem CID: 69036 IUPAC Name: 2-(benzenesulfonyl)ethylsulfonylbenzene SMILES: O=S(=O)(CCS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1