missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylsulfonyl)ethane, 98+%

Catalog No. AAL0408314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0408314 25 g
AAL0408306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0408314 Supplier Thermo Scientific Chemicals Supplier No. L0408314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 599-94-0 | C14H14O4S2 | 310.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 599-94-0
Molecular Formula C14H14O4S2
Molecular Weight (g/mol) 310.38
MDL Number MFCD00041256
InChI Key ULELOBVZIKJPAC-UHFFFAOYSA-N
Synonym 1,2-bis phenylsulfonyl ethane, 1,2-bis phenylsulphonyl ethane, 1,1'-ethane-1,2-diylbis sulphonyl bisbenzene, 2-benzenesulfonyl ethylsulfonylbenzene, ethylenebisphenyl sulfone, 2-phenylsulfonyl ethyl sulfonyl benzene, 2-benzenesulfonyl ethanesulfonyl benzene, 1,1'-ethane-1,2-diyldisulfonyl dibenzene, ethylene diphenylsulfone, acmc-1apw3
PubChem CID 69036
IUPAC Name 2-(benzenesulfonyl)ethylsulfonylbenzene
SMILES O=S(=O)(CCS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 179°C to 180°C
Quantity 25 g
Beilstein 1888062
Formula Weight 310.39
Percent Purity ≥98%
Chemical Name or Material 1,2-Bis(phenylsulfonyl)ethane
EINECSNumber 209-979-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only