CAS RN 59702-31-7
IUPAC Name:
1-ethylpiperazine-2,3-dione
Synonyms:
1-ethylpiperazine-2,3-dione
Molecular Weight (g/mol):
142.16
Molecular Formula:
C6H10N2O2
InChi Key:
ZBEKOEYCWKIMGU-UHFFFAOYSA-N
SMILES:
CCN1CCNC(=O)C1=O
1
–
1
of
1
results
1
1-Ethyl-2,3-dioxopiperazine 98.0+%, TCI America™
CAS: 59702-31-7 Molecular Formula: C6H10N2O2 Molecular Weight (g/mol): 142.158 MDL Number: MFCD00051825 InChI Key: ZBEKOEYCWKIMGU-UHFFFAOYSA-N Synonym: 1-ethyl-2,3-dioxopiperazine,n-ethyl-2,3-dioxopiperazine,1-ethyl-2,3-piperazinedione,2,3-piperazinedione, 1-ethyl,4-ethyl-2,3-dioxopiperazine,4-ethyl-2,3-piperazinedione,ccris 8931,4-ethyl-2,3-dioxo-piperazine,4-ethyl-2,3-dioxypiperazine,n-ethylpiperazine-2,3-dione PubChem CID: 108812 IUPAC Name: 1-ethylpiperazine-2,3-dione SMILES: CCN1CCNC(=O)C1=O