CAS RN 5963-77-9
IUPAC Name:
pent-2-ynoic acid
Synonyms:
pent-2-ynoic acid
Molecular Weight (g/mol):
98.1
Molecular Formula:
C5H6O2
InChi Key:
MINRDQDGBLQBGD-UHFFFAOYSA-N
SMILES:
CCC#CC(O)=O
1
–
1
of
1
results
1
Promotions Available
2-Pentynoic acid, 97%
2-Pentynoic acid, 97%
CAS: 5963-77-9 Molecular Formula: C5H6O2 Molecular Weight (g/mol): 98.101 MDL Number: MFCD00014370 InChI Key: MINRDQDGBLQBGD-UHFFFAOYSA-N Synonym: 2-pentynoic acid,pentynoic acid,2-pentynoicacid,2-pentyneoic acid,acmc-1ay8s PubChem CID: 324379 IUPAC Name: pent-2-ynoic acid SMILES: CCC#CC(=O)O