accessibility menu, dialog, popup

UserName

CAS RN 596-01-0

IUPAC Name: 3,3-bis(4-hydroxynaphthalen-1-yl)-1,3-dihydro-2-benzofuran-1-one
Synonyms: naphtholphthalein α-naphtholphthalein
Molecular Weight (g/mol): 418.45
Molecular Formula: C28H18O4
InChi Key: HQHBAGKIEAOSNM-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=C2C=CC=CC2=C(O)C=C1
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Color 
  • (3)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)