missing translation for 'onlineSavingsMsg'
Learn More

α-Naphtholphthalein, Solstice

Catalog No. 60046240
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046240 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046240 Supplier Solstice Supplier No. 3342410G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 596-01-0
Molecular Formula C28H18O4
Molecular Weight (g/mol) 418.448
MDL Number MFCD00036202
InChI Key HQHBAGKIEAOSNM-UHFFFAOYSA-N
PubChem CID 68993
IUPAC Name 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O

Specifications

Melting Point 238°C to 240°C
Packaging Glass bottle
UN Number NONH for all modes of transport
Beilstein 97816
Color Brown
Quantity 10 g
Physical Form Powder
Formula Weight 418.44g/mol
Chemical Name or Material α-Naphtholphthalein
RTECSNumber 209-875-5