missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Naphtholphthalein, pure, indicator

Catalog No. AC190120250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190120250 25 g Glass bottle
AC190120010 1 g Glass bottle
AC190120050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC190120250 Supplier Thermo Scientific Chemicals Supplier No. 190120250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 596-01-0 | C28H18O4 | 418.448 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 596-01-0
Molecular Formula C28H18O4
Molecular Weight (g/mol) 418.448
MDL Number MFCD00036202
InChI Key HQHBAGKIEAOSNM-UHFFFAOYSA-N
PubChem CID 68993
IUPAC Name 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O

Specifications

Absorbance 550 min. (at lamda max)
Melting Point 240.0°C to 250.0°C
Infrared Spectrum Authentic
Lambda Max 648 to 654nm (in 0.1N NaOH)
Loss on Drying 5% max. (110°C, 1 hr.)
Packaging Glass bottle
Visual Transition Interval from pH 7.3 (pinkish-yellow) to pH 8.7 (greenish--blue)
Beilstein 18, I, 384
Merck Index 15, 6474
Color Beige to Pink or Brown
Quantity 25 g
Physical Form Powder
Formula Weight 418.45
Grade Pure
Chemical Name or Material α-Naphtholphthalein
Show More Show Less
EINECSNumber 209-875-5
TSCA TSCA

RUO – Research Use Only