CAS RN 5949-29-1
Citric Acid Monohydrate (Granular/Certified ACS), Fisher Chemical™
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 MDL Number: MFCD00149972 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric Acid Monohydrate (Granular/USP), Fisher Chemical™
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 MDL Number: MFCD00149972 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric acid monohydrate, 99.5%, for analysis
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 MDL Number: MFCD00149972 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric acid monohydrate, 99+%, ACS reagent
CAS: 5949-29-1 Molecular Formula: C6H8O7·H2O Molecular Weight (g/mol): 210.15 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric acid monohydrate, for HPLC, Thermo Scientific™
Citric acid monohydrate, 99.5+%
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 MDL Number: MFCD00149972 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric acid monohydrate, ACS, 99.0-102.0%
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 MDL Number: MFCD00149972 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric Acid Monohydrate, ACS Reagent, Reagents
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.14 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: 2-hydroxypropane-1,2,3-tricarboxylic Acid monohydrate IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate SMILES: O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid Monohydrate, Solstice
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 MDL Number: MFCD00149972 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric Acid Monohydrate, Purified, Reagents
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.14 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: 2-hydroxypropane-1,2,3-tricarboxylic Acid monohydrate IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate SMILES: O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid Monohydrate, TraceSELECT™, ≥99.9998% (Metals basis), Solstice
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 MDL Number: MFCD00149972 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric Acid, Monohydrate, Granular, USP, 99.5-100.5%, Spectrum™ Chemical
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.14 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate SMILES: O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid, Monohydrate, Granular, FCC, 99.5-100.5%, Spectrum™ Chemical
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.14 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate SMILES: O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid Monohydrate, For Analysis, ACS, EMSURE™, MilliporeSigma™
CAS: 5949-29-1 Molecular Formula: C6H10O8 Molecular Weight (g/mol): 210.138 InChI Key: YASYEJJMZJALEJ-UHFFFAOYSA-N Synonym: citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp PubChem CID: 22230 ChEBI: CHEBI:31404 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Citric Acid, Monohydrate, Crystal, ACS, 99.0-102.0%, Spectrum™ Chemical
CAS: 5949-29-1