Learn More
Citric acid monohydrate, ACS, 99.0-102.0%
Description
Citric acid monohydrate is used in the preparation of citrate buffer in platelets for intravital microscopy. It acts as a pH-control agent in foods, beverages and pharmaceuticals applications. It acts as an iron chelator. In animals, it improves the utilization of nutritional calcium. It is a useful buffer component for antigen and epitope unmasking. It is also used as an acidifier, a flavoring agent and a chelating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Density | 1.542 g/mL |
| Quantity | 2 kg |
| Odor | Odorless |
| Physical Form | Solid |
| Sensitivity | Hygroscopic |
| Merck Index | 14,2326 |
| Solubility Information | Soluble in water,ethanol and ether. Partially soluble in methanol,diethyl ether. Insoluble in n-octanol. |
| Formula Weight | 210.14 (192.13 Anhydrous) |
| Percent Purity | 99.0 to 102.0% |
| Show More |
Safety and Handling
| Hazard Category | H319-H500 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 201-069-1 |
| RTECSNumber | GE7810000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only