missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, ACS Reagent, Reagents

Catalog No. 01338183
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338183 10 kg
01338182 25 kg
01338181 3 kg
01338180 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01338183 Supplier Labchem Inc Supplier No. C110450010KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Citric Acid Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid monohydrate
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153°C
Color Colorless/White
Quantity 10 kg
Packaging Poly Pail
Physical Form Solid
Specific Gravity 1.542
Grade ACS
Chemical Name or Material Citric Acid Monohydrate, ACS Reagent