missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid monohydrate, for HPLC, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | Citric acid monohydrate |
| CAS | 5949-29-1 |
| Formule moléculaire | C6H10O8 |
| Formule linéaire | HO2CCH2C(OH)(CO2H)CH2CO2H·H2O |
| Renseignements sur la solubilité | Solubility aq. soln.: 676 g/L (25°C). Other solubilities: 2.17 g/100 g ether - 0.007 g/100 g chloroform, 15.43 g/100 g amyl alcohol - 5.98 g/100 g amyl acetate, 5.28 g/100 g ethyl acetate - 197 g/100 g methanol, 62.8 g/100 g propanol |
| Clé InChI | YASYEJJMZJALEJ-UHFFFAOYSA-N |
| SOURIRES | O.OC(=O)CC(O)(CC(O)=O)C(O)=O |
| Nom de l’IUPAC | 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate |
| Poids moléculaire (g/mol) | 210.14 |
| Poids de formule | 210.15 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
RUO – Research Use Only