accessibility menu, dialog, popup

UserName

NH2NBrN

CAS RN 59489-71-3

IUPAC Name: 5-bromopyrazin-2-amine
Synonyms: 5-bromopyrazin-2-amine
Molecular Weight (g/mol): 174
Molecular Formula: C4H4BrN3
InChi Key: KRRTXVSBTPCDOS-UHFFFAOYSA-N
SMILES: NC1=CN=C(Br)C=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)