missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyrazine, 97%

Catalog No. AAH2746506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2746506 5 g
AAH2746503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2746506 Supplier Thermo Scientific Chemicals Supplier No. H2746506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59489-71-3 | C4H4BrN3 | 174.001 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59489-71-3
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD00235015
InChI Key KRRTXVSBTPCDOS-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyrazine, 5-bromo-2-pyrazinamine, 5-bromo-pyrazin-2-ylamine, 2-bromo-5-aminopyrazine, pyrazinamine, 5-bromo, 5-amino-2-bromopyrazine, 5-bromo-2-aminopyrazine, 5-bromopyrazin-2-ylamine, 5-bromopyrazinamine, 2-amino-5-bromo-pyrazine
PubChem CID 599539
IUPAC Name 5-bromopyrazin-2-amine
SMILES C1=C(N=CC(=N1)Br)N

Specifications

Melting Point 113°C to 117°C
Quantity 5 g
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromopyrazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only