accessibility menu, dialog, popup

UserName

CAS RN 5943-34-0

IUPAC Name: bis(propan-2-yl)trisulfane
Synonyms: diisopropyltrisulfane
Molecular Weight (g/mol): 182.36
Molecular Formula: C6H14S3
InChi Key: WUZSBMCRYUJOEU-UHFFFAOYSA-N
SMILES: CC(C)SSSC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results