missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Trisulfide 98.0+%, TCI America™

Catalog No. D43005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43005G 5 g
D430025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43005G Supplier TCI America Supplier No. D43005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5943-34-0
Molecular Formula C6H14S3
Molecular Weight (g/mol) 182.36
MDL Number MFCD00040042
InChI Key WUZSBMCRYUJOEU-UHFFFAOYSA-N
PubChem CID 80046
IUPAC Name bis(propan-2-yl)trisulfane
SMILES CC(C)SSSC(C)C

Specifications

Color Yellow
Boiling Point 79°C
Quantity 5 g
Formula Weight 182.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Trisulfide
EINECSNumber (2)-2421
TSCA No