accessibility menu, dialog, popup

UserName

CAS RN 59227-89-3

H3CNO

CAS RN 59227-89-3

IUPAC Name: 1-dodecylazepan-2-one
Synonyms: laurocapram tranzone laurocapramum caswell no. 064A usaf ND-72
Molecular Weight (g/mol): 281.48
Molecular Formula: C18H35NO
InChi Key: AXTGDCSMTYGJND-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN1CCCCCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results