missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laurocapram 96.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -7°C |
| Color | Yellow |
| Boiling Point | 160°C |
| Quantity | 5 mL |
| Formula Weight | 281.48 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Laurocapram |
Sécurité et manipulation
| RTECSNumber | CM3995000 |
| TSCA | No |