accessibility menu, dialog, popup

UserName

CAS RN 591-82-2

IUPAC Name: 1-isothiocyanato-2-methylpropane
Synonyms: 1-isothiocyanato-2-methylpropane
Molecular Weight (g/mol): 115.19
Molecular Formula: C5H9NS
InChi Key: NSDDRJXKROCWRZ-UHFFFAOYSA-N
SMILES: CC(C)CN=C=S
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)