missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl isothiocyanate, 97%

Catalog No. AAA1893214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1893214 25 g
AAA1893206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1893214 Supplier Thermo Scientific Chemicals Supplier No. A1893214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 591-82-2 | C5H9NS | 115.194 g/mol

It is isothiocyanate compound used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is isothiocyanate compound used for proteomics research.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.

Chemical Identifiers

CAS 591-82-2
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.194
MDL Number MFCD00022062
InChI Key NSDDRJXKROCWRZ-UHFFFAOYSA-N
Synonym isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci
PubChem CID 68960
IUPAC Name 1-isothiocyanato-2-methylpropane
SMILES CC(C)CN=C=S

Specifications

Density 0.94
Boiling Point 160°C to 162°C
Flash Point 24°C (75°F)
Odor Pungent
Refractive Index 1.498
Quantity 25 g
UN Number UN2924
Beilstein 1740371
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material Isobutyl isothiocyanate
Show More Show Less
Hazard Category H226-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 209-733-2
RTECSNumber NX8780100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only