missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl isothiocyanate, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 97.0 |
| Density | 0.9400g/mL |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Linear Formula | (CH3)2CHCH2NCS |
| Quantity | 10g |
| Formula Weight | 115.2 |
| Percent Purity | 97% |
| Chemical Name or Material | Isobutyl isothiocyanate |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement Causes serious eye damage. Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. Causes severe skin burns and eye damage. Flammable liquid and vapor. Highly flammable liquid and vapor |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediat |