accessibility menu, dialog, popup

UserName

OHBr

CAS RN 591-20-8

IUPAC Name: 3-bromophenol
Synonyms: M-bromophenol
Molecular Weight (g/mol): 173.01
Molecular Formula: C6H5BrO
InChi Key: MNOJRWOWILAHAV-UHFFFAOYSA-N
SMILES: OC1=CC=CC(Br)=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)