accessibility menu, dialog, popup

UserName

CAS RN 59016-93-2

IUPAC Name: [4-(hydroxymethyl)phenyl]boronic acid
Synonyms: 4-(hydroxymethyl)phenylboronic acid
Molecular Weight (g/mol): 151.96
Molecular Formula: C7H9BO3
InChi Key: PZRPBPMLSSNFOM-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)B(O)O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)