missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H12041G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12041G Supplier TCI America Supplier No. H12041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59016-93-2
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.956
MDL Number MFCD00792672
InChI Key PZRPBPMLSSNFOM-UHFFFAOYSA-N
Synonym 4-hydroxymethyl phenylboronic acid, 4-hydroxymethyl benzeneboronic acid, 4-hydroxymethyl phenyl boronic acid, 4-hydroxymethylphenyl boronic acid, 4-hydroxymethylphenylboronic acid, 4-boronobenzyl alcohol, p-hydroxymethyl phenylboronic acid, 4-hydroxymethyl phenyl boranediol
PubChem CID 2734706
IUPAC Name [4-(hydroxymethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CO)(O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 151.96
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride)