accessibility menu, dialog, popup

UserName

CAS RN 59-48-3

IUPAC Name: 1H-indol-2-ol
Synonyms: oxindole
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: JHFAEUICJHBVHB-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=CC=C2N1
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)