accessibility menu, dialog, popup

UserName

H3CCH3(R)HNO(S)OH(R)NH2OHO

CAS RN 58970-76-6

IUPAC Name: (2R)-2-[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanamido]-4-methylpentanoic acid
Synonyms: bestatin
Molecular Weight (g/mol): 308.38
Molecular Formula: C16H24N2O4
InChi Key: VGGGPCQERPFHOB-MCIONIFRSA-N
SMILES: CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)