accessibility menu, dialog, popup

UserName

HOONH

CAS RN 5893-05-0

IUPAC Name: 2-[(triphenylmethyl)amino]acetic acid
Synonyms: [(triphenylmethyl)amino]acetic acid
Molecular Weight (g/mol): 317.39
Molecular Formula: C21H19NO2
InChi Key: XUXRXWLKUYPGMZ-UHFFFAOYSA-N
SMILES: OC(=O)CNC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results