missing translation for 'onlineSavingsMsg'
Learn More

N-(Triphenylmethyl)glycine 98.0+%, TCI America™

Catalog No. T13065G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13065G Supplier TCI America Supplier No. T13065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5893-05-0
Molecular Formula C21H19NO2
Molecular Weight (g/mol) 317.39
MDL Number MFCD00004276,MFCD07776921
InChI Key XUXRXWLKUYPGMZ-UHFFFAOYSA-N
Synonym n-tritylglycine, n-triphenylmethyl glycine, trt-gly-oh, trityl-glycine, 2-tritylamino acetic acid, glycine, n-triphenylmethyl, 2-triphenylmethyl amino acetic acid, triphenylmethyl amino acetic acid, glycine,n-trityl, tritylamino acetic acid
PubChem CID 248898
IUPAC Name 2-[(triphenylmethyl)amino]acetic acid
SMILES OC(=O)CNC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 317.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(Triphenylmethyl)glycine