accessibility menu, dialog, popup

UserName

CAS RN 589-81-1

IUPAC Name: 3-methylheptane
Synonyms: 3-methylheptane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: LAIUFBWHERIJIH-UHFFFAOYNA-N
SMILES: CCCCC(C)CC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)