accessibility menu, dialog, popup

UserName

HON

CAS RN 586-95-8

IUPAC Name: (pyridin-4-yl)methanol
Synonyms: 4-pyridinemethanol
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: PTMBWNZJOQBTBK-UHFFFAOYSA-N
SMILES: OCC1=CC=NC=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)