missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinemethanol, 99%

Catalog No. AAA1469836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1469836 500 g
AAA1469814 25 g
AAA1469822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1469836 Supplier Thermo Scientific Chemicals Supplier No. A1469836
Only null left

CAS: 586-95-8 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 586-95-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006442
InChI Key PTMBWNZJOQBTBK-UHFFFAOYSA-N
Synonym 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol
PubChem CID 11472
SMILES OCC1=CC=NC=C1

Specifications

Melting Point 53°C to 55°C
Boiling Point 141°C (12 mmHg)
Flash Point 148°C (298°F)
Quantity 500 g
Beilstein 107850
Sensitivity Hygroscopic
IUPAC Name (pyridin-4-yl)methanol
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 4-Pyridinemethanol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-590-6
RTECSNumber UT4691000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only