accessibility menu, dialog, popup

UserName

CAS RN 58438-03-2

IUPAC Name: (2S)-2-amino-3-(naphthalen-2-yl)propanoic acid
Synonyms: β-(2-naphthyl)-alanine
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: JPZXHKDZASGCLU-LBPRGKRZSA-N
SMILES: N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)