accessibility menu, dialog, popup

UserName

CAS RN 58207-19-5

IUPAC Name: N-{2-chloro-1-[3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl}-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride
Synonyms: clindamycin hydrochloride [usan:ban:jan] hydrochloride
Molecular Weight (g/mol): 461.44
Molecular Formula: C18H34Cl2N2O5S
InChi Key: AUODDLQVRAJAJM-UHFFFAOYNA-N
SMILES: Cl.CCCC1CC(N(C)C1)C(=O)NC(C(C)Cl)C1OC(SC)C(O)C(O)C1O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Color 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)