missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin hydrochloride monohydrate

Catalog No. AAJ6140906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6140906 5 g
AAJ6140903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6140906 Supplier Thermo Scientific Chemicals Supplier No. J6140906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clindamycin hydrochloride, CAS # 21462-39-5, is the salt of clindamycin, a derivative of the broad-spectrum bacteriostatic antibiotic lincomycin. | CAS: 58207-19-5 | C18H34Cl2N2O5S | 461.439 g/mol

Clindamycin hydrochloride monohydrate is an antibacterial agent that is a semisynthetic analog of lincomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Clindamycin is a lincosamide antibiotic that disrupts protein synthesis by interfering with the aminoacyl-tRNA ribosome complex

• Clindamycin decreases sebaceous gland activity and blocks the neuromuscular junction

Application

• Clindamycin is commonly used in laboratory applications to inhibit the growth anaerobic microbes, but has some effectiveness against aerobic microbes as well

• It can be used in the laboratory study of cell culture for resistance testing, or in antibiotic allergic reaction studies

Chemical Identifiers

CAS 58207-19-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-NFOLQJLNSA-N
PubChem CID 131632848
IUPAC Name (2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Type Clindamycin Hydrochloride Monohydrate
Physical Form Powder
Color White
Quantity 5 g
Beilstein 4070786
Merck Index 14,2356
Solubility Information Soluble in water
Formula Weight 479.46 (461.44 Anhydrous)
Chemical Name or Material Clindamycin hydrochloride monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-398-6
RTECSNumber GF2275000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only