missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014748
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014748 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014748 Supplier Merck Emd Millipore Supplier No. PHR11591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Clindamycin is a common antibiotic drug generally employed for the treatment of infections caused by gram-positive aerobes and both gram-negative and gram-positive anaerobic pathogens. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 58207-19-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C18H33 ClN2O5 S · HCl · H2O
Synonym Clindamycin hydrochloride monohydrate; (7 S)-7-Chloro-7-deoxylincomycin hydrochloride monohydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 479.46
Grade Certified Reference Material