accessibility menu, dialog, popup

UserName

KOO

CAS RN 582-25-2

IUPAC Name: potassium benzoate
Synonyms: potassium benzoate
Molecular Weight (g/mol): 160.21
Molecular Formula: C7H5KO2
InChi Key: XAEFZNCEHLXOMS-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (3)
Percent Purity 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (7)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)